Learn More
Sterol carrier protein 2 Antibody (SR2036), Novus Biologicals™
Código de producto. p-200005611 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18669734 100 μg 100 microlitros
18689863 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18669734 Proveedor Novus Biologicals N.º de proveedor NBP322523100ul
Rabbit Monoclonal Antibody
Sterol carrier protein 2 Monoclonal antibody specifically detects Sterol carrier protein 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Sterol carrier protein 2
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence
Clasificación Monoclonal
Clon SR2036
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulación PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Alias de gen DKFZp686C12188, DKFZp686D11188, NLTP, non-specific lipid-transfer protein, NSL-TP, Propanoyl-CoA C-acyltransferase, SCP-2, SCP-CHI, SCPX, SCP-X, sterol carrier protein 2EC 2.3.1.176, Sterol carrier protein X
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human Sterol carrier protein 2 (Uniprot #: P22307)
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6342
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20° C. Avoid freeze/thaw cycles.
Formulario Purified
Isotype IgG