missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
STEAP1 Monoclonal antibody specifically detects STEAP1 in Human samples. It is validated for Flow Cytometry,Western Blot
Especificaciones
Especificaciones
| Antígeno | STEAP1 |
| Aplicaciones | Flow Cytometry, Western Blot |
| Clasificación | Monoclonal |
| Clon | 1057323 |
| Conjugado | Alexa Fluor 532 |
| Dilución | Western Blot, Flow Cytometry |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | EC 1.16.1.-, PRSS24MGC19484, six transmembrane epithelial antigen of the prostate, six transmembrane epithelial antigen of the prostate 1, Six-transmembrane epithelial antigen of prostate 1, STEAPmetalloreductase STEAP1 |
| Especie del huésped | Mouse |
| Inmunógeno | E. coli-derived recombinant human STEAP-1, Met1-Trp71, Accession # Q9UHE8 |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?