missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT1 Mouse anti-Human, Mouse, Rat, Clone: 1F7C6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66545-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene belongs to the SMC3 subfamily of SMC proteins. The encoded protein occurs in certain cell types as either an intracellular, nuclear protein or a secreted protein. The nuclear form, known as structural maintece of chromosomes 3, is a component of the multimeric cohesin complex that holds together sister chromatids during mitosis, enabling proper chromosome segregation. Post-translational modification of the encoded protein by the addition of chondroitin sulfate chains gives rise to the secreted proteoglycan bamacan, an abundant basement membrane protein.Especificaciones
| STAT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P42224, P42225 | |
| STAT1 | |
| STAT1 Fusion Protein Ag0199 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1F7C6 | |
| Unconjugated | |
| STAT1 | |
| ISGF 3, STAT1, STAT91 | |
| Mouse | |
| Protein A | |
| RUO | |
| 20846, 25124, 6772 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido