missing translation for 'onlineSavingsMsg'
Learn More

START domain containing 7 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Código de producto. 18098229 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18098229 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18098229 Supplier Novus Biologicals Supplier No. NBP297964AF594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

START domain containing 7 Polyclonal specifically detects START domain containing 7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno START domain containing 7
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Alexa Fluor 594
Dilución Immunohistochemistry, Immunohistochemistry-Paraffin
Formulación 50mM Sodium Borate
Alias de gen Gestational trophoblastic tumor protein 1, GTT1stAR-related lipid transfer protein 7, mitochondrial, StARD7, StAR-related lipid transfer (START) domain containing 7, START domain containing 7, START domain-containing protein 7
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human START domain containing 7 fragment.
Método de purificación Antigen Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 56910
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.