missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Staphylococcus Glu-C Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
1 mg
250 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
250 microgramos
50 microgramos
500 microgramos
Descripción
Recombinant Staphylococcus Glu-C is a 28.8 kDa protease consisting of 266 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, as they are frequently used in the analysis and production of proteins. Glu-C cleaves at the Carboxyl side of E (can also cleave D under certain conditions).
Especificaciones
Especificaciones
| Para utilizar con (aplicación) | Functional Assay |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 66839245 |
| Peso molecular | 28.8 kDa |
| Nombre | Staphylococcus Glu-C |
| Cantidad | 50 μg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Nombre común | Glu-C |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido