missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ STAMBP Recombinant Protein

Human STAMBP full-length ORF ( AAH65574, 1 a.a. - 424 a.a.) recombinant protein with GST-tag at N-terminal

Marca:  Abnova™ H00010617-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16165432

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Cytokine-mediated signal transduction in the JAK-STAT cascade requires the involvement of adaptor molecules. One such signal-transducing adaptor molecule contains an SH3 domain that is required for induction of MYC and cell growth. The protein encoded by this gene binds to the SH3 domain of the signal-transducing adaptor molecule, and plays a critical role in cytokine-mediated signaling for MYC induction and cell cycle progression. Multiple alternatively spliced transcript variants encoding the same protein isoform have been found for this gene. (provided by RefSeq)

  • Molecular weight: 72.38kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8 in the elution buffer
  • Quality Control Testing:12.5% SDS-PAGE stained with Coomassie Blue

Best use within three months from the date of receipt of this protein

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Especificaciones

Especificaciones

AAH65574
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
72.38
8
Glutathione Sepharose 4 Fast Flow
25 ug
MSDHGDVSLPPEDRVRALSQLGSAVEVNEDIPPRRYFRSGVEIIRMASIYSEEGNIEHAFILYNKYITLFIEKLPKHRDYKSAVIPEKKDTVKKLKEIAFPKAEELKAELLKRYTKEYTEYNEEKKKEAEELARNMAIQQELEKEKQRVAQQKQQQLEQEQFHAFEEMIRNQELEKERLKIVQEFGKVDPGLGGPLVPDLEKPSLDVFPTLTVSSIQPSDCHTTVRPAKPPVVDRSLKPGALSNSESIPTIDGLRHVVVPGRLCPQFLQLASANTARGVETCGILCGKLMRNEFTITHVLIPKQSAGSDYCNTENEEELFLIQDQQGLITLGWIHTHPTQTAFLSSVDLHTHCSYQMMLPESVAIVCSPKFQETGFFKLTDHGLEEISSCRQKGFHPHSKDPPLFCSCSHVTVVDRAVTITDLR
AMSH/MGC126516/MGC126518
STAMBP
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10617
STAMBP (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
STAMBP
Human
Recombinant
Solution
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales