Learn More
Esferas de acero inoxidable, 2 mm de diámetro, tipo 302, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar 042016.KM
Detalles adicionales : Peso : 0.01000kg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Stainless Steel spheres, Type 302 | |
Fe:Cr:Ni:Mn:Si; 70:18:9:2:1 wt% | |
25 Pc. |
2mm (0.08 in.) dia. | |
MFCD00148499 |