missing translation for 'onlineSavingsMsg'
Learn More

ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A Antibody, Novus Biologicals™

Código de producto. p-200045372 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
25 μL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18415291 25 μL 25 microlitros
18219487 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18415291 Proveedor Novus Biologicals N.º de proveedor NBP18704525ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A Polyclonal specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A
Aplicaciones Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gen Alpha 2,3-ST 1, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1, DKFZp666E036, DKFZp779K2051, EC 2.4.99, EC 2.4.99.4, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4A, sialyltransferase 4A (beta-galactosidase alpha-2,3-sialytransferase), sialyltransferase 4A (beta-galactoside alpha-2,3-sialyltransferase), sialyltransferase 4A (beta-galactoside alpha-2,3-sialytransferase), SIAT4, SIAT4A, SIAT4-A, SIATFLFLJ36548, ST3 beta-galactoside alpha-2,3-sialyltransferase 1, ST3Gal I, ST3GalA.1MGC9183, ST3GalI, ST3GalIA, ST3GalIA1,Beta-galactoside alpha-2,3-sialyltransferase 1, ST3OST3GalA
Símbolos de los genes ST3GAL1
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids:MVLELSENLKRLIKHRPCTCTHCIGQRKLSAWFDERFNQTMQPLLTAQNALLEDDTYRWWLRLQREKKPNNLNDTIKELFRVVPGNVDPMLEKRSVGCRRCAVVGNSGNLRESSYGPEIDSHDFVLRMNKAPTAGFEA
Método de purificación Affinity Purified
Cantidad 25 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6482
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.