missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SSRP1 Antibody, Novus Biologicals™

Código de producto. 30067174 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Unit Size:
100 microgramos
25 microgramos
Product Code. Cantidad unitSize
30067174 100 μg 100 microgramos
30067103 25 μg 25 microgramos
2 options
This item is not returnable. View return policy

Product Code. 30067174

Brand: Novus Biologicals™ NBP330846100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SSRP1 Polyclonal antibody specifically detects SSRP1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno SSRP1
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot : 5-10 μg/mL, Immunoprecipitation
Formulación Phosphate buffered saline
Alias de gen Chromatin-specific transcription elongation factor 80 kDa subunit, cisplatin-DNA SSRP, facilitates chromatin remodeling 80 kDa subunit, Facilitates chromatin transcription complex 80 kDa subunit, Facilitates chromatin transcription complex subunit SSRP1, FACT, FACT complex subunit SSRP1, FACT80FACT 80 kDa subunit, FACTp80, high mobility group box, hSSRP1, Recombination signal sequence recognition protein 1, structure specific recognition protein 1, Structure-specific recognition protein 1, T160
Especie del huésped Rabbit
Inmunógeno Full-length recombinant SSRP1 protein tagged with histidine.
Método de purificación Ammonium sulfate precipitation
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6749
Especies diana Human, Mouse
Contenido y almacenamiento Store at -20°C. Avoid freeze/thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.