Learn More
SSEA-1 Antibody (MC-480), Novus Biologicals™
Código de producto. 18009223 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
18009223 0.025 mL 25 microlitros
18775853 0.1 mL 0.10 ml
2 options
Código de producto. 18009223
Marca: Novus Biologicals NB1001831SS
Mouse Monoclonal Antibody has been used in 7 publications
SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Mouse, Rat, Chimpanzee samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno SSEA-1
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon MC-480
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 2 ug/ml, Simple Western 1:10, Flow Cytometry 2.5 ug/million cells, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200
Formulación PBS with 0.02% Sodium Azide
Alias de gen alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Símbolos de los genes FUT4
Especie del huésped Mouse
Inmunógeno F9 tetracarcinoma stem cells (X-irradiated).
Método de purificación IgM purified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells
Primario o secundario Primary
ID de gen (Entrez) 2526
Especificidad de la prueba Human and mouse. This antibody recognizes SSEA1 in mouse EC and ES cells but not human, but SSEA1 is expressed by some human differentiated cell types (ie: trophoblasts). Reactivity with Rat is reported by Kim et al. 2007 (PMID 17548530).
Especies diana Human, Mouse, Rat, Chimpanzee
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgM
Mostrar más Mostrar menos
For Research Use Only