Learn More
SPZ1 Antibody, Novus Biologicals™
Código de producto. 18140582 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
18140582 0.025 mg 25 microgramos
18717283 0.1 mg 0.10 miligramo
2 options
Código de producto. 18140582
Marca: Novus Biologicals NBP224582SS
Rabbit Polyclonal Antibody
SPZ1 Polyclonal specifically detects SPZ1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno SPZ1
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulación 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
N.º de referencia del gen Q9BXG8
Alias de gen FLJ25709, NYD-TSP1, spermatogenic leucine zipper 1, spermatogenic leucine zipper protein 1, spermatogenic zip 1, Testis-specific protein 1, Testis-specific protein NYD-TSP1, TSP1
Símbolos de los genes SPZ1
Especie del huésped Rabbit
Inmunógeno A portion of amino acids 40-90 of human SPZ1 was used as the immunogen.
Método de purificación Affinity Purified
Cantidad 0.025 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 84654
Especificidad de la prueba The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in rat, mouse, monkey and dog, and 82% homologous in chicken.
Especies diana Human, Mouse, Rat, Primate
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only