missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
SPRED1 Polyclonal antibody specifically detects SPRED1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | SPRED1 |
| Aplicaciones | ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | FITC |
| Formulación | PBS |
| Alias de gen | EVH1/Sprouty domain containing protein, FLJ33903, hSpred1, NFLS, spred-1, sprouty-related, EVH1 domain containing 1, sprouty-related, EVH1 domain-containing protein 1, suppressor of Ras/MAPK activation |
| Especie del huésped | Rabbit |
| Inmunógeno | SPRED1 antibody was raised against a 14 amino acid synthetic peptide near the center of the human SPRED1. The immunogen is located within amino acids 250 - 300 of SPRED1. Amino Acid Squence: RHPDMWKNDLERDD |
| Método de purificación | Peptide affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?