missing translation for 'onlineSavingsMsg'
Learn More

Splicing Factor 1 Antibody (OTI7H9) - Azide and BSA Free, Novus Biologicals™

Código de producto. 18127616 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
18127616 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18127616

Marca: Novus Biologicals NBP274338

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Splicing Factor 1 Monoclonal specifically detects Splicing Factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Splicing Factor 1
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon OTI7H9
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1:2000, Immunohistochemistry 1:150
Formulación Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Alias de gen Mammalian branch point-binding protein, mBBP, splicing factor 1, Transcription factor ZFM1, ZFM1BBP, Zinc finger gene in MEN1 locus, Zinc finger protein 162D11S636, ZNF162MBBP
Símbolos de los genes SF1
Especie del huésped Mouse
Inmunógeno Human recombinant protein fragment corresponding to amino acids 1-276 of human SF1 (NP_973727) produced in E.coli.
Método de purificación Affinity Purified
Cantidad 100 μg
Primario o secundario Primary
ID de gen (Entrez) 7536
Reconstitución Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Especies diana Human
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.