missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24661-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
SPINK9 is a serine protease inhibitor which specifically inhibits KLK5. It may contribute to the regulation of the desquamation process in skin by inhibiting KLK5.Especificaciones
SPINK9 | |
Polyclonal | |
Unconjugated | |
SPINK9 | |
LEKTI2, SPINK9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
643394 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5DT21 | |
SPINK9 | |
SPINK9 Fusion Protein Ag19924 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |