missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antígeno | Spike RBD |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 1042425 |
| Conjugado | PE-Cyanine7 |
| Dilución | Flow Cytometry |
| Formulación | PBS |
| Especie del huésped | Mouse |
| Inmunógeno | Chinese Hamster Ovary cell line CHO-derived SARS-CoV Spike RBD, Arg306-Phe527, Accession # NP_0828851.1 |
| Método de purificación | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?