Learn More
SPATA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 11849-1-AP-20UL
 Ver más versiones de este producto
Código de producto. 16859193
  • 145.00€ / 20 microlitros
Descripción
May play a role in spermatid maturation or sperm function.
Especificaciones
Especificaciones
SPATA6
Polyclonal
Unconjugated
SPATA6
HASH, SPATA6, spermatogenesis associated 6, SRF 1, SRF1
Rabbit
Antigen Affinity Chromatography
RUO
171413, 54558, 67946
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3U6K5, Q99MU5, Q9NWH7
Spata6
SPATA6 Fusion Protein Ag2413
20 μL
Primary
Human, Mouse, Rat