Descripción
SOX9 Polyclonal specifically detects SOX9 in Human, Mouse, Rat, Canine, Feline, Goat, Primate, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
| Antígeno | SOX9 |
| Aplicaciones | Western Blot |
| Especies diana | Human, Canine, Goat, Primate |
For Research Use Only