missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SOX11 Polyclonal specifically detects SOX11 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antígeno | SOX11 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay |
| Clasificación | Polyclonal |
| Concentración | 0.2mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500, Chromatin Immunoprecipitation (ChIP) |
| Formulación | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Alias de gen | SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11 |
| Símbolos de los genes | SOX11 |
| Especie del huésped | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?