missing translation for 'onlineSavingsMsg'
Learn More

SOX10 Mouse anti-Human, Mouse, Clone: SOX10/991, Novus Biologicals™

Código de producto. p-7121236 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18208634 20 μg 20 microgramos
18246833 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18208634 Proveedor Novus Biologicals N.º de proveedor NBP25962020UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

SOX10 Monoclonal specifically detects SOX10 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, Flow (Intracellular).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SOX10
Aplicaciones Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
Clasificación Monoclonal
Clon SOX10/991
Concentración 0.2 mg/mL
Conjugado Unconjugated
Dilución Western Blot 0.5-1 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml, SDS-Page, Protein Array 1:100-1:2000
Formulación 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Alias de gen MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of
Símbolos de los genes SOX10
Especie del huésped Mouse
Inmunógeno Recombinant human SOX10 protein fragment (around aa115-269) (exact sequence is proprietar
Peso molecular del antígeno 54 kDa
Método de purificación Protein A or G purified
Cantidad 20 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6663
Especificidad de la prueba The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. Recognizes a protein of ∼55kDa, identified as SOX10. This MAb is highly specific and does not cross-react with other members of the SOX-family. SOX genes comprise a family of genes that are related to the mammalian sex-determining gene SRY. These genes similarly contain sequences that encode for the HMG -box domain, which is responsible for the sequence-specific DNA-binding activity. SOX-10 is a sensitive marker of melanoma, including conventional, spindled, and desmoplastic subtypes. It is expressed by metastatic melanomas and nodal capsular nevus in sentinel lymph nodes, but not by other lymph node components such as dendritic cells, which usually express S100 protein. Commonly used melanoma markers, such as anti-HMB-45 and anti-Melan-A, are poorly expressed in desmoplastic melanomas while SOX-10 is moderately to strongly expressed in desmoplastic melanomas. SOX-10 is considered as a very reliable marker for recognizing residual desmoplastic melanomas. In normal tissues, it is expressed in Schwann cells, melanocytes, and myoepithelial cells of salivary, bronchial and mammary glands. SOX-10 expression is also observed in mast cells
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2b κ
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.