missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R1/SSTR1 Antibody - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Somatostatin R1/SSTR1 Polyclonal antibody specifically detects Somatostatin R1/SSTR1 in Human, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | Somatostatin R1/SSTR1 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | G-protein coupled receptor, somatostatin receptor 1, somatostatin receptor type 1, SRIF-2, SS1R, SS-1-R, SS1-R, SSR1 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 1-56 of human Somatostatin R1/SSTR1 (NP_001040.1). MFPNGTASSPSSSPSPSPGSCGEGGGSRGPGAGAADGMEEPGRNASQNGTLSEGQG |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?