Learn More
SNX21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22193-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
May be involved in several stages of intracellular trafficking.Especificaciones
| SNX21 | |
| Polyclonal | |
| Unconjugated | |
| SNX21 | |
| C20orf161, PP3993, SNX L, SNX21, SNXL, Sorting nexin 21, sorting nexin family member 21, Sorting nexin L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 90203 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q969T3 | |
| SNX21 | |
| SNX21 Fusion Protein Ag16900 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |