missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SNRP70 Monoclonal antibody specifically detects SNRP70 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | SNRP70 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL |
| Formulación | PBS (pH 7.3), 50% glycerol, 0.05% BSA |
| Alias de gen | RNPU1ZU170K, RPU1U1AP, small nuclear ribonucleoprotein 70kDa (U1), Snp1, snRNP70, SNRP70U1 small nuclear ribonucleoprotein 70 kDa, U1 snRNP 70 kDa, U1-70Ksmall nuclear ribonucleoprotein 70kDa (RNP antigen), U1AP1, U1RNP |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthetic peptide corresponding to a sequence within amino acids 201-300 of SNRP70 (NP_003080.2).,, Sequence:, RGGADVNIRHSGRDDTSRYDERPGPSPLPHRDRDRDRERERRERSRERDKERERRRSRSRDRRRRSRSRDKEERRRSRERSKDKDRDRKRRSSRSRERAR |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?