Learn More
SMC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556150
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: KLTQEETNFK SLVHDLFQKV EEAKSSLAMN RSRGKVLDAI IQEKKSGRIP GIYGRLGDLG AIDEKYDVAI SSCCHALDYI VVDSIDIAQE CVNFLKRQNI GVATFIGLDK MAVWAKKMTE IQTPENTPRL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.Especificaciones
SMC4 | |
Polyclonal | |
Unconjugated | |
Smc4 | |
2500002A22Rik; C79747; Capc; CAP-C; chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4; SMC-4; SMC4 structural maintenance of chromosomes 4-like 1; SMC4L1; structural maintenance of chromosomes 4; structural maintenance of chromosomes protein 4; XCAP-C homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10051, 295107, 70099 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8CG47, Q9NTJ3 | |
Smc4 | |
Recombinant protein corresponding to Human SMC4 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only