missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad9 [p Tyr92] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-33580-20ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Smad9 Monoclonal antibody specifically detects Smad9 in Human samples. It is validated for ELISA,Western Blot
Especificaciones
| Smad9 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| MAD homolog 9, MAD, mothers against decapentaplegic homolog 9 (Drosophila), Madh6, MADH6mothers against decapentaplegic homolog 9, MADH9Mothers against decapentaplegic, drosophila, homolog of, 9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9SMAD8B, SMAD, mothers against DPP homolog 9, SMAD, mothers against DPP homolog 9 (Drosophila), SMAD8, SMAD8A, Smad9 | |
| A synthetic phosphorylated peptide around Y92 of human Smad9.1). | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4093 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido