Learn More
Smad4 Antibody (CL9774) [Janelia Fluor™ 646], Novus Biologicals™
Código de producto. 30298130 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30298130 0.1 mL 0.10 ml
1 options
Código de producto. 30298130 Proveedor Novus Biologicals N.º de proveedor NBP305507JF646
Mouse Monoclonal Antibody
Smad4 Monoclonal antibody specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Smad4
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL9774
Conjugado Janelia Fluor 646
Formulación 50mM Sodium Borate
Alias de gen DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from Q13485, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction, Stem Cell Signaling Pathway
Primario o secundario Primary
ID de gen (Entrez) 4089
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1