missing translation for 'onlineSavingsMsg'
Learn More

Smad3, p Ser423, p Ser425 Antibody, Novus Biologicals™

Código de producto. 18791464 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
Tamaño de la unidad:
25 microgramos
Código de producto. Cantidad unitSize
18791464 0.025 mg 25 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18791464

Marca: Novus Biologicals NBP177836SS

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Smad3 (p Ser423, p Ser425) Polyclonal antibody specifically detects Smad3 (p Ser423, p Ser425) in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Single Cell Western.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Smad3 (p Ser423, p Ser425)
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 1.1 mg/ml
Conjugado Unconjugated
Dilución Western Blot 1:2000-1:20000, Flow Cytometry 1:10-1:1000, ELISA 1:15000-1:30000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Single Cell Western 100 ug/ml
Formulación 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
N.º de referencia del gen NC_000015.9
Alias de gen DKFZp586N0721, DKFZp686J10186, hMAD-3, hSMAD3, HsT17436, JV15-2MGC60396, MAD homolog 3, mad protein homolog, MAD, mothers against decapentaplegic homolog 3, MAD, mothers against decapentaplegic homolog 3 (Drosophila), Mad3, MADH3mad homolog JV15-2, mothers against decapentaplegic homolog 3, Mothers against DPP homolog 3, pSMAD, SMA- and MAD-related protein 3, SMAD 3, SMAD family member 3HSPC193, SMAD, mothers against DPP homolog 3, SMAD, mothers against DPP homolog 3 (Drosophila), Smad3
Símbolos de los genes SMAD3
Especie del huésped Rabbit
Inmunógeno Smad3 [p Ser423, p Ser425] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a dual phosphorylated synthetic peptide corresponding to a c-terminal region with Serine 423 and Serine 425 of human Smad3 protein. (Uniprot: P84022)
Peso molecular del antígeno 48.08 kDa
Método de purificación Affinity Purified
Cantidad 0.025 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4088
Especificidad de la prueba This affinity-purified antibody is directed against the phosphorylated form of human Smad3 protein at the pS423 and pS425 residues. The resultant affinity purified antibody was then cross adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against human Smad3 pS423 and pS425 protein and the antibody is specific for the phosphorylated form of the protein. Reactivity with non-phosphorylated human Smad3 is minimal by ELISA and western blot. Expect reactivity against phosphorylated Smad1 and Smad5. Negligible reactivity is seen against other phosphorylated Smad family members. A BLAST analysis was used to suggest cross reactivity with Smad3 from human, Xenopus species laevis, Xenopus species tropicalis, zebrafish, rat, mouse, swine, bovine and chicken based on 100% sequence homology with the immunogen. Reactivity against homologues from other sources is not known.
Especies diana Human, Mouse, Rat, Pig, Bovine, Chicken, Xenopus species, Zebrafish
Contenido y almacenamiento Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.