Learn More
Slow Skeletal Muscle Myosin Heavy Chain Antibody (M14), Janelia Fluor™ 646, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Slow Skeletal Muscle Myosin Heavy Chain Monoclonal specifically detects Slow Skeletal Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
| Antígeno | Slow Skeletal Muscle Myosin Heavy Chain |
| Aplicaciones | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | M14 |
| Conjugado | Janelia Fluor 646 |
| Dilución | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Especie del huésped | Mouse |
| Inmunógeno | purified human skeletal muscle myosin |
| Método de purificación | Protein A purified |
| Mostrar más |