Learn More
SLC6A2/NET/Noradrenaline transporter Antibody, Novus Biologicals™
Código de producto. 18007705 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
18007705 100 μL 100 microlitros
1 options
Código de producto. 18007705
Marca: Novus Biologicals NBP160120
Rabbit Polyclonal Antibody
SLC6A2/NET/Noradrenaline transporter Polyclonal antibody specifically detects SLC6A2/NET/Noradrenaline transporter in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno SLC6A2/NET/Noradrenaline transporter
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/ml
Conjugado Unconjugated
Dilución Western Blot 1.0 ug/ml
Formulación PBS, 2% Sucrose with 0.09% Sodium Azide
N.º de referencia del gen P23975
Alias de gen NAT1neurotransmitter transporter, NET, NET1sodium-dependent noradrenaline transporter, Norepinephrine transporter, SLC6A5, solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2, Solute carrier family 6 member 2, solute carrier family 6 member 5
Símbolos de los genes SLC6A2
Especie del huésped Rabbit
Inmunógeno Synthetic peptides corresponding to SLC6A2(solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2) The peptide sequence was selected from the middle region of SLC6A2. Peptide sequence STLSGSTFWAVVFFVMLLALGLDSSMGGMEAVITGLADDFQVLKRHRKLF The peptide sequence for this immunogen was taken from within the described region.
Método de purificación Affinity purified
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6530
Especificidad de la prueba Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Chicken: 78%.
Reconstitución Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Especies diana Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only
Título del producto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.