missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A2/NET/Noradrenaline transporter Antibody - Azide and BSA Free, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
SLC6A2/NET/Noradrenaline transporter Polyclonal antibody specifically detects SLC6A2/NET/Noradrenaline transporter in Human, Mouse, Rat samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | SLC6A2/NET/Noradrenaline transporter |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500 - 1:1000 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | NAT1neurotransmitter transporter, NET, NET1sodium-dependent noradrenaline transporter, Norepinephrine transporter, SLC6A5, solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2, Solute carrier family 6 member 2, solute carrier family 6 member 5 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 30-235 of human SLC6A2/NET/Noradrenaline transporter (NP_478066.3). ELIPDAPLSSAAYSIRSIGERPVLKAPVPKRQKCDHWTPCPSDTYAYRLLSGGGRSKYAKICFEDNLLMGEQLGNVARGINIAIVNYVTGNVTATRCFDMYEGDNSGPMTKFIQSAAPKSLLFMVTYDDGSTRLNNDAKNAIEALGSKEIRNMKFRSSWVFIAAKGLELPSEIQREKINHSDAKNNRYSGWPAEIQIEGCIPKERS |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?