missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
SLC25A41 Polyclonal antibody specifically detects SLC25A41 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | SLC25A41 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | DyLight 350 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | FLJ40442, MGC34725, solute carrier family 25 member 41, solute carrier family 25, member 41 |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with E. coli-derived Human SLC25A41 fragment. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?