missing translation for 'onlineSavingsMsg'
Learn More

SLC13A1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18589523 Shop All Bio Techne Products
Change view
Click to view available options
:
0.1 mL, DyLight 680
Product Code.
18589523 0.1 mL, DyLight 680
1 options
This item is not returnable. View return policy

Product Code. 18589523

Brand: Novus Biologicals NBP298609FR

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC13A1 Polyclonal antibody specifically detects SLC13A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno SLC13A1
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado DyLight 680
Formulación 50 mM Sodium Borate
Alias de gen solute carrier family 13 member 1, NAS1, NaSi-1, solute carrier family 13 (sodium/sulfate symporters), member 1
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human SLC13A1 fragment.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6561
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.