missing translation for 'onlineSavingsMsg'
Learn More

FGF10 Recombinant Human Protein, Invitrogen™ Sino Biological™

Código de producto. 15216955
Change view
Click to view available options
Cantidad:
5 x 50 μg
50 μg
5 μg
Tamaño de la unidad:
250 microgramos
5 microgramos
50 microgramos
Código de producto. Cantidad unitSize
15216955 5 x 50 μg 250 microgramos
15226955 5 μg 5 microgramos
15236955 50 μg 50 microgramos
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15216955

Marca: Invitrogen 10573HNAE250

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln 38-Ser 208)

Fibroblast Growth Factor 10 (FGF10), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

TRUSTED_SUSTAINABILITY

Especificaciones

Para utilizar con (aplicación) Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
ID de gen (Entrez) 2255
Peso molecular 19.6kDa
Método de purificación PAGE
Cantidad 5 x 50 μg
Requisitos de almacenamiento Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Forma estructural Full Length/Recombinant
Alias de gen FGF10
Nombre común Fibroblast Growth Factor 10
Símbolo de gen FGF10
Etiqueta de proteína Untagged
Sistema de expresión E. coli
Familia de proteínas Growth Factors & Receptors
Formulario Lyophilized
Grado de pureza o calidad ≥90% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.