missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Siglec-7/CD328 Monoclonal antibody specifically detects Siglec-7/CD328 in Human samples. It is validated for Flow Cytometry, CyTOF-reported
Especificaciones
Especificaciones
| Antígeno | Siglec-7/CD328 |
| Aplicaciones | Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 194211 |
| Conjugado | PerCP |
| Dilución | Flow Cytometry, CyTOF-reported |
| Formulación | PBS |
| Alias de gen | Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7 |
| Especie del huésped | Mouse |
| Inmunógeno | Mouse myeloma cell line NS0-derived recombinant human Siglec-7, Gln19-Gly357 (predicted), Accession # Q9Y286 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?