Learn More
Eurofins DiscoverX SHP-2 Protein
Código de producto. 17360444
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 μg
250 μg
Tamaño de la unidad:
10 microgramos
250 microgramos
17360444 10 μg 10 microgramos
17370444 250 μg 250 microgramos
2 options
Código de producto. 17360444
Marca: Eurofins DiscoverX 14622
N-terminal GST-tagged recombinant, human SHP-2 amino acids 230-545, for use in Phosphatase Assays.
SHP-2 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains two tandem Src homology-2 domains, which function as phospho-tyrosine binding domains and mediate the interaction of this PTP with its substrates. This PTP is widely expressed in most tissues and plays a regulatory role in various cell signaling events that are important for a diversity of cell functions, such as mitogenic activation, metabolic control, transcription regulation, and cell migration. Mutations in this gene are a cause of Noonan syndrome as well as acute myeloid leukemia.
GenBank X70766
Especificaciones
Formulación mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 10% glycerol, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Peso molecular 63.7 kDa
Cantidad 10 μg
Especie Human
Recombinante Recombinant
Etiqueta de proteína N-terminal GST
Sistema de expresión Recombinant enzyme expressed in E. coli cells
Forma de proteína Truncated
Grado de pureza o calidad ≥76%
Expresión SHP-2, active