Learn More
Eurofins DiscoverX SHP-2 Protein
Código de producto. p-7228126
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 μg
250 μg
Tamaño de la unidad:
10 microgramos
250 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17360444 10 μg 10 microgramos
17370444 250 μg 250 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17360444 Proveedor Eurofins DiscoverX N.º de proveedor 14622
N-terminal GST-tagged recombinant, human SHP-2 amino acids 230-545, for use in Phosphatase Assays.
SHP-2 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains two tandem Src homology-2 domains, which function as phospho-tyrosine binding domains and mediate the interaction of this PTP with its substrates. This PTP is widely expressed in most tissues and plays a regulatory role in various cell signaling events that are important for a diversity of cell functions, such as mitogenic activation, metabolic control, transcription regulation, and cell migration. Mutations in this gene are a cause of Noonan syndrome as well as acute myeloid leukemia.
GenBank X70766
Especificaciones
Formulación mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 10% glycerol, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Peso molecular 63.7 kDa
Cantidad 10 μg
Especie Human
Recombinante Recombinant
Etiqueta de proteína N-terminal GST
Sistema de expresión Recombinant enzyme expressed in E. coli cells
Forma de proteína Truncated
Grado de pureza o calidad ≥76%
Expresión SHP-2, active