Learn More
Eurofins DiscoverX SHP-1 Protein
Descripción
SHP-1 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. N-terminal part of this PTP contains two tandem Src homolog (SH2) domains, which act as protein phospho-tyrosine binding domains, and mediate the interaction of this PTP with its substrates. This PTP is expressed primarily in hematopoietic cells, and functions as an important regulator of multiple signaling pathways in hematopoietic cells. This PTP has been shown to interact with, and dephosphorylate a wide spectrum of phospho-proteins involved in hematopoietic cell signaling. Multiple alternatively spliced variants of this gene, which encode distinct isoforms, have been reported.
GenBank NM_002831
Especificaciones
Especificaciones
| Formulación | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 10% glycerol, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Peso molecular | 95 kDa |
| Cantidad | 10 μg |
| Especie | Human |
| Recombinante | Recombinant |
| Etiqueta de proteína | N-terminal GST |
| Sistema de expresión | Recombinant enzyme expressed in E. coli cells |
| Forma de proteína | Full-length |
| Grado de pureza o calidad | ≥54% |
| Expresión | SHP-1, active |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.