Learn More
Abnova™ SHH (Human) Recombinant Protein (Q01)
Descripción
This gene encodes a protein that is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites. Of three human proteins showing sequence and functional similarity to the sonic hedgehog protein of Drosophila, this protein is the most similar. The protein is made as a precursor that is autocatalytically cleaved; the N-terminal portion is soluble and contains the signalling activity while the C-terminal portion is involved in precursor processing.
- Theoretical MW: 36.74kDa
- Preparation Method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Best use within three months from the date of receipt of this protein
Enzyme Linked Immunosorbent Assay, Western Blotting, Antibody Production, Protein Array
Especificaciones
Especificaciones
| Número de acceso | NP_000184 |
| Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulación | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| ID de gen (Entrez) | 6469 |
| Peso molecular | 36.74 |
| Nombre | SHH (Human) Recombinant Protein (Q01) |
| Intervalo de pH | 8 |
| Método de preparación | In vitro wheat germ expression system |
| Método de purificación | Glutathione Sepharose 4 Fast Flow |
| Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.