missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ Varian SPI Liner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Marca: Trajan™ 09202701
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Varian/Bruker instruments
Especificaciones
0.25 mm Restriction | |
54 mm | |
0.5 mm | |
Varian GC Instruments |
1 Ea. | |
SPI Liner | |
4.6 mm |