missing translation for 'onlineSavingsMsg'
Learn More

SEZ6/BSRP-C Antibody (Abbv-011), CoraFluor™ 1, Novus Biologicals™

Código de producto. 30144973 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30144973 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30144973 Supplier Novus Biologicals Supplier No. NBP328127CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Human Monoclonal Antibody

SEZ6/BSRP-C Monoclonal antibody specifically detects SEZ6/BSRP-C in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antígeno SEZ6/BSRP-C
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon Abbv-011
Conjugado CoraFluor 1
Formulación PBS
Alias de gen hSEZ-6, seizure related 6 homolog (mouse)
Especie del huésped Human
Inmunógeno SEZ6
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 124925
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.