missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SESN3 Polyclonal antibody specifically detects SESN3 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | SESN3 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Formulación | PBS (pH 7.4) |
| N.º de referencia del gen | AAH17296.1 |
| Alias de gen | SEST3MGC29667, sestrin 3, sestrin-3 |
| Especie del huésped | Rabbit |
| Inmunógeno | SESN3 (AAH17296.1, 1 a.a. - 321 a.a.) full-length human protein. MNRGGGSPSAAANYLLCTNCRKVLRKDKRIRVSQPLTRGPSAFIPEKEVVQANTVDERTNFLVEEYSTSGRLDNITQVMSLHTQYLESFLRSQFYMLRMDGPLPLPYRHYIAIMAAARHQCSYLINMHVDEFLKTGGIAEWLNGLEYVPQRLKNLNEINKLLAHRPWLITKEHIQKLVKTGENNWSLPELVHAVVLLAHYHALASFVFGSGINPERDPEISNGFRLISVNNFCVCDLANDNNIENASLSGSNFGIVDSLSELEALMERMKRLQEEREDEEASQEEMSTRFEKEKKESLFVVSGDTFHSFPHSGAFLHFFAF |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?