Learn More
Abnova™ SERPINF1 (Human) Recombinant Protein
Código de producto. 16168911
Click to view available options
Cantidad:
10 μg
25 μg
missing translation for 'unitSize'
10 microgramos
25 microgramos
This item is not returnable. View return policy
Product Code. 16168911
missing translation for 'mfr': Abnova™ H00005176P01.25ug
This item is not returnable. View return policy
Human SERPINF1 full-length ORF ( AAH00522.1, 17 a.a. - 418 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: SSCQNPASPPEEGSPDPDSTGALVEEEDPFFKVPVNKLAAAVSNFGYDLYRVRSSTSPTTNVLLSPLSVATALSALSLGAEQRTESIIHRALYYDLISSPDIHGTYKELLDTVTAPQKNLKSASRIVFEKKLRIKSSFVAPLEKSYGTRPRVLTGNPRLDLQEINNWVQAQMKGKLARSTKEIPDEISILLLGVAHFKGQWVTKFDSRKTSLEDFYLDEERTVRVPMMSDPKAVLRYGLDSDLSCKIAQLPLTGSMSIIFFLPLKVTQNLTLIEESLTSEFIHDIDRELKTVQAVLTVPKLKLSYEGEVTKSLQEMKLQSLFDSPDFSKITGKPIKLTQVEHRAGFEWNEDGAGTTPSPGLQPAHLTFPLDYHLNQPFIFVLRDTDTGALLFIGKILDPRGP
Specifications
Número de acceso AAH00522.1
ID de gen (Entrez) 5176
Nombre serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1
Método de preparación Wheat germ expression system
Pruebas de control de calidad 125% SDS-PAGE Stained with Coomassie Blue
Cantidad 25 μg
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Alias de gen EPC-1, PEDF, PIG35
Símbolo de gen SERPINF1
Especie Wheat Germ (in vitro)
Etiqueta de proteína GST
Tampón 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.