missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Serine racemase Monoclonal specifically detects Serine racemase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
| Antígeno | Serine racemase |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | OTI2E3 |
| Conjugado | DyLight 350 |
| Dilución | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | D-serine ammonia-lyase, D-serine dehydratase, EC 4.3.1.17, EC 4.3.1.18, EC 5.1.1.18, ILV1, ISO1, L-serine ammonia-lyase, L-serine dehydratase, serine racemase |
| Símbolos de los genes | SRR |
| Especie del huésped | Mouse |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?