Learn More
Secretogranin II Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20357-1-AP-150UL
Descripción
The Secretogranin II antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Secretogranin II. This antibody recognizes human, mouse, rat antigen. The Secretogranin II antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
| Secretogranin II | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 7857 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA | |
| 233 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |