Learn More
Secretogranin II Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. p-7224043 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16879884 20 μL 20 microlitros
16869884 150 μL 150 microlitros
2 options
Código de producto. 16879884
Marca: Proteintech 203571AP20UL
Rabbit Polyclonal Antibody
The Secretogranin II antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Secretogranin II. This antibody recognizes human, mouse, rat antigen. The Secretogranin II antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Antígeno Secretogranin II
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA
Clasificación Polyclonal
Concentración 233 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 7857
Especies diana Human, Rat, Mouse
Isotype IgG