Learn More
Invitrogen™ Human SBK1, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This SBK1 recombinant human protein (full length) was expressed in insect cells. SBK1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for SBK1 is NP_001019572.1.
Specifications
Specifications
| Número de acceso | Q52WX2 |
| Concentración | See Label |
| Para utilizar con (aplicación) | Kinase Assay |
| Formulación | 50mM tris HCl with 10mM glutathione, 150mM NaCl, 0.1mM PMSF, 0.1mM EDTA, 25% glycerol, 0.25mM DTT and no preservative; pH 7.5 |
| ID de gen (Entrez) | 388228 |
| Peso molecular | 72.5 kDa |
| Nombre | Human SBK1, GST Tag |
| Intervalo de pH | 7.5 |
| Método de purificación | Purified |
| Cantidad | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.