missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike S1 Subunit Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Marca: R&D Systems MAB105407-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry
Especificaciones
| SARS-CoV-2 Spike S1 Protein | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| SARS-CoV-2 | |
| E. coli-derived recombinant human SARS-CoV-2 Spike S1 Subunit, Arg319-Phe541, Accession # QHD43416 | |
| 100 μg | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 2811A | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| QHD43416 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido