missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 NSP16 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB10979-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NSP16 Monoclonal antibody specifically detects NSP16 in SARS-CoV-2 samples. It is validated for Western Blot, Western Blot
Especificaciones
| NSP16 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 1044327 | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL | |
| YP_009725311.1 and YP_00972530 | |
| E. coli-derived SARS-CoV-2 NSP16/NSP10 protein, (Ser1-Asn298) and (Ala1-Gln139), Accession # YP_009725311.1 and YP_00972530 | |
| 25 μg | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido