Learn More
SARS-CoV-2 Membrane Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
205.00€ - 443.00€
Especificaciones
Antígeno Membrane
Clon 1041508
Dilución Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Aplicaciones Immunohistochemistry, Immunocytochemistry
Clasificación Monoclonal
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
18763207
GREENER_CHOICE
Ver documentos
R&D Systems
MAB10690-SP
25 μg
205.00€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
18761437
GREENER_CHOICE
Ver documentos
R&D Systems
MAB10690-100
100 μg
443.00€
100 microgramos
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
Membrane Monoclonal antibody specifically detects Membrane in SARS-CoV-2 samples. It is validated for Immunohistochemistry, Immunocytochemistry
Especificaciones
Especificaciones
Membrane
Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
E. coli-derived SARS-CoV-2 membrane (M) protein, Met101-His222, Accession # YP_009724369
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1041508
Immunohistochemistry, Immunocytochemistry
Unconjugated
Mouse
SARS-CoV-2
YP_009724369
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.