missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
SARM1 Polyclonal antibody specifically detects SARM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Especie del huésped | Rabbit |
| Método de purificación | Peptide affinity purified |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 23098 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?