Descripción
- 3-level coding for safe sample and vessel identification
- Codes on three sides of plates for flexible reading
- ECC 200 error correction function enables safe code reading with up to 30% damaged codes
- Additional, ID-specific information on plates available via Eppendorf™ DataPort for documentation purposes
- Direct import into eLabInventory and eLabJournal software for convenient handling
- Import to other database systems possible
- Manufactured from high-quality polypropylene for highest sample purity
Especificaciones
Especificaciones
| Código de barras | Includes Barcode |
| Formato | Estándar SBS |
| matriz, red, conjunto | 8 x 12 |
| Línea de productos | Eppendorf |
| Color | Clear |
| Tipo de producto | Microplaca |
| Material | Polypropylene |
| Volumen del pocillo (métrico) | 320 μl |
| N.° de pocillos | 96 |
| Esterilidad | Estéril |
| Mostrar más |